ENA258390139 Enamine

[6-(1H-1,3-benzodiazol-1-yl)pyridin-3-yl]methanamine

  • MDL number MFCD09734576
  • Cas Number : 953748-84-0
  • Molecular Weight : 224.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-59116

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59116
Availability details for
Quantity
  • Availability
  • Add to Cart