ENA258390178 Enamine

8-oxatricyclo[7.4.0.0,2,7]trideca-1(9),2(7),3,5,10,12-hexaen-5-amine hydrochloride

  • MDL number MFCD00136984
  • Cas Number : 91493-29-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59291

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59291
Availability details for
Quantity
  • Availability
  • Add to Cart