ENA282547880 Enamine

2,2,2-trifluoroethyl N-(4-phenyl-1,3-thiazol-2-yl)carbamate

  • MDL number MFCD15209532
  • Cas Number : 1235438-81-9
  • Molecular Weight : 302.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-56011

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56011
Availability details for
Quantity
  • Availability
  • Add to Cart