ENA282547885 Enamine

2,2,2-trifluoroethyl N-[5-(dimethylsulfamoyl)-2-methoxyphenyl]carbamate

  • MDL number MFCD15209537
  • Cas Number : 1235440-73-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56634

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56634
Availability details for
Quantity
  • Availability
  • Add to Cart