ENA282547886 Enamine

2,2,2-trifluoroethyl N-[2-(4-methoxyphenoxy)pyridin-3-yl]carbamate

  • MDL number MFCD15209538
  • Cas Number : 1221725-36-5
  • Molecular Weight : 342.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,2,2-trifluoroethyl 2-(4-methoxyphenoxy)pyridin-3-ylcarbamate
Inchi Code 1S/C15H13F3N2O4/c1-22-10-4-6-11(7-5-10)24-13-12(3-2-8-19-13)20-14(21)23-9-15(16,17)18/h2-8H,9H2,1H3,(H,20,21)
Inchi Key HSXWXVXREIACDR-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-56773

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56773
Availability details for
Quantity
  • Availability
  • Add to Cart