ENA282547887 Enamine

phenyl N-(3-aminophenyl)carbamate

  • MDL number MFCD15209539
  • Cas Number : 68621-59-0
  • Molecular Weight : 228.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56852

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-56852
Availability details for
Quantity
  • Availability
  • Add to Cart