ENA282547889 Enamine

3-oxo-6-phenyl-1H,2H,3H-pyrazolo[3,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD15209541
  • Cas Number : 1221724-64-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-56939

Safety Information

msds https://www.enaminestore.com/catalog/EN300-56939
Availability details for
Quantity
  • Availability
  • Add to Cart