ENA282547893 Enamine

4-(4-fluorophenyl)-1-methyl-1H-imidazol-5-amine

  • MDL number MFCD15209546
  • Cas Number : 1221725-63-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59314

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59314
Availability details for
Quantity
  • Availability
  • Add to Cart