ENA282547895 Enamine

1-[4-(3,4-difluorophenoxy)-2-(trifluoromethyl)phenyl]methanamine hydrochloride

  • MDL number MFCD15209548
  • Cas Number : 1221725-70-7
  • Molecular Weight : 339.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [4-(3,4-difluorophenoxy)-2-(trifluoromethyl)phenyl]methanamine hydrochloride
Inchi Code 1S/C14H10F5NO.ClH/c15-12-4-3-10(6-13(12)16)21-9-2-1-8(7-20)11(5-9)14(17,18)19;/h1-6H,7,20H2;1H
Inchi Key AELMIEIGACJDMX-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-59492

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59492
Availability details for
Quantity
  • Availability
  • Add to Cart