ENA282547896 Enamine

tert-butyl 3-(1-aminoethyl)piperidine-1-carboxylate

  • MDL number MFCD15209549
  • Cas Number : 1235439-55-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59555

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59555
Availability details for
Quantity
  • Availability
  • Add to Cart