ENA282547900 Enamine

1,2-dimethyl-1H-1,3-benzodiazole-5-carboxylic acid hydrochloride

  • MDL number MFCD15209554
  • Cas Number : 1221724-84-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 304...306
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-59914

Safety Information

msds https://www.enaminestore.com/catalog/EN300-59914
Availability details for
Quantity
  • Availability
  • Add to Cart