ENA282547904 Enamine

2-{[(pyridin-2-yl)methyl]amino}acetic acid dihydrochloride

  • MDL number MFCD15209558
  • Cas Number : 820220-92-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60171

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60171
Availability details for
Quantity
  • Availability
  • Add to Cart