ENA282547906 Enamine

sodium 1,3-thiazole-5-carboxylate

  • MDL number MFCD15209560
  • Cas Number : 26862-80-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Na+
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60199

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60199
Availability details for
Quantity
  • Availability
  • Add to Cart