ENA282547911 Enamine

1-[3-(aminomethyl)phenyl]-N-methylmethanesulfonamide hydrochloride

  • MDL number MFCD15209566
  • Cas Number : 1221723-71-2
  • Molecular Weight : 250.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-60293

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60293
Availability details for
Quantity
  • Availability
  • Add to Cart