ENA282547913 Enamine

[1-phenyl-3-(pyridin-4-yl)-1H-pyrazol-4-yl]methanamine

  • MDL number MFCD15209572
  • Cas Number : 1221723-97-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60369

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60369
Availability details for
Quantity
  • Availability
  • Add to Cart