ENA282547915 Enamine

2,2,2-trifluoroethyl N-[1-(4-methyl-4H-1,2,4-triazol-3-yl)ethyl]carbamate

  • MDL number MFCD15209574
  • Cas Number : 1221725-80-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60386

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60386
Availability details for
Quantity
  • Availability
  • Add to Cart