ENA282547917 Enamine

3-(1,4-diazepane-1-carbonyl)-1H-indole hydrochloride

  • MDL number MFCD15209576
  • Cas Number : 1235439-36-7
  • Molecular Weight : 279.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60444

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60444
Availability details for
Quantity
  • Availability
  • Add to Cart