ENA282547920 Enamine

2-[(naphthalen-2-yl)methyl]pyrrolidine hydrochloride

  • MDL number MFCD15209581
  • Cas Number : 1221726-17-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-60486

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60486
Availability details for
Quantity
  • Availability
  • Add to Cart