ENA282547921 Enamine

6-amino-5-chloropyridine-3-carboxamide

  • MDL number MFCD15209582
  • Cas Number : 1221724-36-2
  • Molecular Weight : 171.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-60511

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60511
Availability details for
Quantity
  • Availability
  • Add to Cart