ENA282547923 Enamine

(2-methyl-5-nitrophenyl)hydrazine dihydrochloride

  • MDL number MFCD15209584
  • Cas Number : 1221725-20-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 189...191
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60525

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60525
Availability details for
Quantity
  • Availability
  • Add to Cart