ENA282547924 Enamine

1-[1-(3-methoxyphenyl)-1H-1,2,3-triazol-4-yl]methanamine hydrochloride

  • MDL number MFCD15209585
  • Cas Number : 1235441-33-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60528

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60528
Availability details for
Quantity
  • Availability
  • Add to Cart