ENA282547925 Enamine

methyl 6-oxo-1,6-dihydropyridine-3-carboxylate hydrochloride

  • MDL number MFCD15209586
  • Cas Number : 1221722-65-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169...171
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60531

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60531
Availability details for
Quantity
  • Availability
  • Add to Cart