ENA282547930 Enamine

5-chloro-1H-[1,2,3]triazolo[4,5-b]pyridine

  • MDL number MFCD09261014
  • Cas Number : 73895-37-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 208-210
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60552

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60552
Availability details for
Quantity
  • Availability
  • Add to Cart