ENA282547934 Enamine

N-(2-chloro-4-fluorophenyl)-2-(N-hydroxyimino)acetamide

  • MDL number MFCD15209595
  • Cas Number : 259860-02-1
  • Molecular Weight : 216.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-60566

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60566
Availability details for
Quantity
  • Availability
  • Add to Cart