ENA282547936 Enamine

5-bromo-1-(propan-2-yloxy)-2,3-dihydro-1H-indene

  • MDL number MFCD15209597
  • Cas Number : 1221725-34-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60584

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60584
Availability details for
Quantity
  • Availability
  • Add to Cart