ENA282547937 Enamine

N1-{5-methyl-[1,2,4]triazolo[1,5-a]pyrimidin-7-yl}ethane-1,2-diamine dihydrochloride

  • MDL number MFCD15209598
  • Cas Number : 1235441-22-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60620

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60620
Availability details for
Quantity
  • Availability
  • Add to Cart