ENA282547938 Enamine

2-benzyl-1,2,3,4-tetrahydroisoquinoline-3-carbohydrazide

  • MDL number MFCD15209599
  • Cas Number : 1251924-04-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60621

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60621
Availability details for
Quantity
  • Availability
  • Add to Cart