ENA282547947 Enamine

2-{[2-(4-bromophenyl)-1,3-thiazol-4-yl]sulfanyl}acetic acid

  • MDL number MFCD15209608
  • Cas Number : 1221726-20-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60657

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60657
Availability details for
Quantity
  • Availability
  • Add to Cart