ENA282547948 Enamine

3-chloro-4-(1H-imidazol-1-yl)aniline hydrochloride

  • MDL number MFCD15209609
  • Cas Number : 1235439-78-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 259...261
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60677

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60677
Availability details for
Quantity
  • Availability
  • Add to Cart