ENA282547949 Enamine

6-(difluoromethoxy)-3,4-dihydronaphthalene-2-sulfonyl chloride

  • MDL number MFCD15209610
  • Cas Number : 1235439-11-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 72...74
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60679

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60679
Availability details for
Quantity
  • Availability
  • Add to Cart