ENA282547952 Enamine

4-imino-3,4-dihydro-1H-2lambda6,3-benzothiazine-2,2-dione

  • MDL number MFCD15209615
  • Cas Number : 27350-16-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60691

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60691
Availability details for
Quantity
  • Availability
  • Add to Cart