ENA282547953 Enamine

1-[(4-chlorophenyl)methyl]-5-hydroxy-3-methyl-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD15209616
  • Cas Number : 1221722-66-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-60692

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60692
Availability details for
Quantity
  • Availability
  • Add to Cart