ENA282547954 Enamine

7-methyl-1,7-diazaspiro[3.5]nonane dihydrochloride

  • MDL number MFCD15209617
  • Cas Number : 1221723-89-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60708

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60708
Availability details for
Quantity
  • Availability
  • Add to Cart