ENA282547955 Enamine

[3-(difluoromethoxy)phenyl]methanamine hydrochloride

  • MDL number MFCD15209619
  • Cas Number : 1106917-71-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-60725

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60725
Availability details for
Quantity
  • Availability
  • Add to Cart