ENA282547956 Enamine

1-(piperidin-4-yl)pyrrolidine-2-carboxamide dihydrochloride

  • MDL number MFCD15209620
  • Cas Number : 864291-96-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60727

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60727
Availability details for
Quantity
  • Availability
  • Add to Cart