ENA282547957 Enamine

methyl[(2-phenyloxan-3-yl)methyl]amine

  • MDL number MFCD15209621
  • Cas Number : 1221724-01-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60734

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60734
Availability details for
Quantity
  • Availability
  • Add to Cart