ENA282547958 Enamine

13-oxo-12-oxa-2,3,7-triazatricyclo[7.4.0.0,2,6]trideca-1(9),3,5,7-tetraene-5-carboxamide

  • MDL number MFCD15209622
  • Cas Number : 1221722-40-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60742

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60742
Availability details for
Quantity
  • Availability
  • Add to Cart