ENA282547959 Enamine

2-(4-bromo-2-hydroxyphenyl)-2-oxoacetic acid

  • MDL number MFCD15209623
  • Cas Number : 1221724-46-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-60743

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60743
Availability details for
Quantity
  • Availability
  • Add to Cart