ENA282547960 Enamine

N-cyclopropylpiperidine-3-carboxamide hydrochloride

  • MDL number MFCD15209624
  • Cas Number : 1221724-04-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148...150
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60748

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60748
Availability details for
Quantity
  • Availability
  • Add to Cart