ENA282547963 Enamine

2-amino-N-(prop-2-yn-1-yl)acetamide hydrochloride

  • MDL number MFCD15209627
  • Cas Number : 1221722-25-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175...177
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60761

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-60761
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart