ENA282547964 Enamine

6-chloro-5-oxo-5,6,7,8-tetrahydronaphthalene-2-sulfonyl chloride

  • MDL number MFCD15209628
  • Cas Number : 1221724-06-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-60772
Availability details for
Quantity
  • Availability
  • Add to Cart