ENA282547966 Enamine

N-(pentan-2-yl)piperidine-3-carboxamide hydrochloride

  • MDL number MFCD15209630
  • Cas Number : 1235440-21-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form oil
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60781

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60781
Availability details for
Quantity
  • Availability
  • Add to Cart