ENA282547967 Enamine

2-methylpentan-2-amine hydrochloride

  • MDL number MFCD00102055
  • Cas Number : 112306-54-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 204...206
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60782

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-60782
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart