ENA282547972 Enamine

3-amino-2-(4-chlorophenyl)-1,1,1-trifluoropropan-2-ol hydrochloride

  • MDL number MFCD15209635
  • Cas Number : 1221722-26-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60809

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60809
Availability details for
Quantity
  • Availability
  • Add to Cart