ENA282547973 Enamine

2-[3-(aminomethyl)phenoxy]-N-cyclopropylacetamide hydrochloride

  • MDL number MFCD15209636
  • Cas Number : 1221723-43-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60843

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60843
Availability details for
Quantity
  • Availability
  • Add to Cart