ENA282547979 Enamine

1-(3-bromophenyl)-5-(trifluoromethyl)-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD15209642
  • Cas Number : 1221725-66-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-60858

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60858
Availability details for
Quantity
  • Availability
  • Add to Cart