ENA282547982 Enamine

1-(4-bromophenyl)-5-(trifluoromethyl)-1H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD15209646
  • Cas Number : 1221724-13-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60866

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60866
Availability details for
Quantity
  • Availability
  • Add to Cart