ENA282547986 Enamine

2-trifluoromethanesulfonylpyridine-3-carboxylic acid

  • MDL number MFCD15209652
  • Cas Number : 1221722-71-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60922

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60922
Availability details for
Quantity
  • Availability
  • Add to Cart