ENA282547987 Enamine

2-(aminomethyl)-N,N-dimethylaniline dihydrochloride

  • MDL number MFCD15209653
  • Cas Number : 1221725-71-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-60937

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60937
Availability details for
Quantity
  • Availability
  • Add to Cart