ENA282547989 Enamine

2-chloro-N-{3-[(thiomorpholin-4-yl)methyl]phenyl}acetamide hydrochloride

  • MDL number MFCD15209656
  • Cas Number : 1221723-47-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-60947

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-60947
Availability details for
Quantity
  • Availability
  • Add to Cart